Advanced Bacteriology Slide Set
Advanced Bacteriology Slide Set, 16, Monera
470183-442
: Avantor
Illustrates the Major Sections Outlined in the 10th Edition of Bergey’s Manual of Systematic Bacteriology
- Identifying features clearly distinguishable
- Rigorous quality control standards
Our set includes 16 slides conveying major sections of the textbook.
: Set includes Section 1: Spirochaetes, Section 2: Aerobic/Microaerobic Motile, Section 4: Gram (-) Aerobic Rods and Cocci, Section 5: Facultatively Anaerobic Gram (-) Rods, Section 8: Anaerobic Gram (-) Cocci, Section 12: Gram (+) Cocci, Section 13: Endospore-Forming Gram (+) Rods and Cocci, Section 15: Irregular, Non-Sporing Gram (+) Rods, and Section 16: Mycobacteria.
- Type:Monera
- Désignation:Advanced Bacteriology Slide Set
- No. of Slides:16