Adjustable Micro Bunsen Burners
Adjustable micro Bunsen burners, Natural gas, 800 - 1200 BTU/cu. ft.
Catalog # 470148-840
Supplier: Shiv Dial Sud & Sons
Fournisseur #: CH10623
Jump to:
Product Details & Documents
Ideal for Jobs Requiring a Small, Intense Flame
- Nickel-plate, die-cast zinc alloy base
- Flame stabilizer and needle valve for precise flame control
- Gas inlet is serrated for secure connections
Gas inlet 5/16" outer dia.; mixing tube 7/16" outer dia.; 3 7/8"H
Spécifications
- Désignation:Adjustable micro Bunsen burners
- For:Natural gas
- BTU Output:800 - 1200 BTU/cu. ft.