Adjustable Bunsen Burners | Avantor
Adjustable Bunsen Burners
Adjustable Bunsen Burners
Certificates
Adjustable Bunsen Burners
Adjustable Bunsen Burners
1 Options Available Below
Product Details & Documents
Precise Flame Control.
  • Gas Inlet: 5/16" O.D.
  • Mixing Tube: 1/2" O.D.
  • Height: 63/8"
These are similar to the standard burner, but with a fine pitch needle valve for flame adjustment and aluminum mixing tube with flame stabilizer.
Product Family Options
Informations sur le produit
  • Désignation
    For
    BTU Output
    Availability
    Price
  • Burner
    Natural gas
    800 - 1200 BTU/cu. ft.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $34.55
    $34.55
    Liquidation
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $345.75
    $345.75
    Liquidation
    Each
       $34.55
    Case of 10
       $345.75
    Supplier Number: CH10620
    Spécifications:
    More
    Specifications
    Cat. No.
    470149-518
  • Gas burner
    LPG
    2000 - 3200 BTU/cu. ft.
    Discontinued
    Supplier Number: C-1484
    Spécifications:
    More
    Specifications
    Cat. No.
    470148-690