Acrylic Buret
Acrylic Buret
Certificates
Acrylic Buret
2 Options Available Below
Product Details & Documents
These hardy burets are ideal for classroom use.
  • Acrylic
  • Transparent
  • Leakproof Stopcock Included
  • Burets Unaffected by Most Dilute Aqueous Solutions
  • Heavy-Duty
The transparent, acrylic burets are supplied with self-lubricating, leakproof PMP stopcock.
They are unaffected by most dilute aqueous solutions except ammonia and HF and are not recommended for use with ethanol or organic solvents.
Product Family Options
Informations sur le produit
  • Capacité
    Intervalle de graduation
    Availability
    Price
  • 100 ml
    0.2 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $193.70
    $193.70
    Each
       $193.70
  • 50 ml
    0.1 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $79.70
    $79.70
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $711.25
    $711.25
    Each
       $79.70
    Case of 10
       $711.25