Justrite® Sure-Grip® EX Acid/Corrosives Steel Safety Cabinet
Fournisseur: Justrite
Certificats
À propos de cet article
Exclusively Formulated Paint and Poly Trays Resist Acids, Bases and Solvents
- 18-gauge double-wall steel construction
- Dual Vents
- Grounding wire connections
- Adjust shelf and leak-proof sill
Additionally, they include polyethylene trays attached to galvanized steel shelves and a separate polyethylene liner for the bottom sump resist aggressive chemicals. Easily remove the liner for quick cleaning of drips and leaks. This popular 30-gallon (114-L) cabinet also includes an extra polyethylene work tray that can be secured to cabinet top for a handy work surface. An all epoxy baked-on powder-coat finish, inside and out, provides increased chemical resistance.
Ordering information: Ten year manufacturer warranty.
Caution: WARNING - This product can expose you to chemicals including lead, which is known to the State of California to cause cancer and birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Frequently Bought Together