Acid Spill Clean Up Kit
470127-676
- Description:Acid Spill Clean Up Kit
- Cat. no.:470127-676
- Supplier No.:IS5003
Ready to Use for Quick Response.
- Handy compact design
- Contains everything to safely contain the spill
Spills are inevitable in almost every chemistry lab. Be prepared for potentially dangerous acid spills with this complete kit that includes everything you need for safe cleanup and disposal. Includes one bag of Vermiculite absorbent, one bag of neutralizing mixture, two vinyl exam gloves, two 9" x 16"polybags, two blank shipping tags, twist ties, and a dust pan and brush set. A detailed instruction sheet/SDS is provided.