Acceleration Trolley
Catalog # 470149-004
Supplier: Shiv Dial Sud & Sons
Specifications
- Description:Acceleration Trolley
- Cat. no.:470149-004
- Supplier No.:P10515A
Specifications
About this item
Show your students how the rate of acceleration of an object depends on the angle of incline with this great hands-on activity.
- Simply fasten to your blackboard or wall and string a cable across the classroom at any angle
Mount the trolley on the cable, add weights with holes provided for weight hangers, and use a stopwatch to measure the time it takes for the trolley to cross.
Includes: trolley with pulley, 9.1m (30 ft.) wire cable, attachment kit with hardware, and instructions.