About this item
These anatomically correct human torsos are a classroom staple.
- Multi-Part Torsos
- Hand-Painted Details
- High-Quality Plastic Construction
- Unisex
- CD-ROM Included
- Includes 3B Smart Anatomy
Each Unisex Torso contains a variety of removable parts. The 12-Part Torso includes a 2-part head, 2-part removable heart, 2 lungs, stomach, liver with gall bladder, 2-part intestinal tract, and front half of a kidney. The 14-Part Torso includes a 3-part head, 2 lungs, 2-part heart, stomach, liver with gall bladder, 2-part intestinal tract, front half of a kidney, and front half of the bladder. The 16-Part Torso includes a 3-part head, 2 lungs with sternum and rib attachments, 2-part heart, stomach, liver with gall bladder, 4-part intestinal tract, front half of kidney, and front half of the bladder. The 21-Part Open Back includes a removable 7th thoracic vertebra, 6-part head, 2-part stomach, lungs, 2-part heart, liver with gall bladder, 4-part intestine, kidney half, bladder half, open back, and brain.
The detailed unisex torso is hand painted and made of high-quality plastic.
Each torso is shipped with the 3B Torso Guide on CD-ROM, which includes keys in multiple languages.