12" Rigid Plastic Ruler
Metric/imperial ruler, Plastic, 305 mm (12")
Catalog # 470177-220
Supplier: Skywill Far East Industries
: 4213
Jump to:
Product Details & Documents
Ideal for classroom use.
- Raised divisions are sharp and clear
- 1/16" divisions on one edge
- 1 mm divisions on the other edge
- Grooved
- Aligned with GLOBE protocols
Spécifications
- Désignation:Metric/imperial ruler
- Longueur:305 mm (12")
- Matière:Plastic