Clear 360° Goniometer
360° Goniometer, 12"
Catalog # 470019-280
Supplier: Fabrication Enterprises
Fournisseur #: 12-1000
Jump to:
Product Details & Documents
Clear Plastic Permits Easy Observation
This goniometer is marked in 1° increments and has three scales calibrated to be used with the International Standards of Measurement. One arm has a linear scale in both inches and centimeters to allow accurate measurement of the joint’s axis and range of motion.
Spécifications
- Désignation:360° Goniometer
- Taille:12"