SP Bel-Art Sterileware® Bent Handle Sampling Lab Scoops, Bel-Art Products, a part of SP
89218-258
- Sterility:Sterile
- Capacity:60 ml (2 oz.)
- Color:White
- Cat. no.:89218-258
- Bowl Dimensions (W×L×H):7.8×4.1×2.9 cm (3¹⁄₁₆×1⅝×1⅛")
- Handle Length:11.6 cm (4⁹⁄₁₆")
- L×W×H:19.4 × 4.1 × 2.9 cm (7⅝ × 1⅝ × 1⅛")
- Supplier No.:H369022006
The curved handle design of the Scienceware® Sterileware® Bent Handle Scoops positions the hand further from the sample when compared to straight handle scoops.
- Durable polystyrene plastic construction
- Sterilized via gamma irradiation
- Lot stamped for accurate lot tracing
- Flat bottom keeps scoops
This reduces the possibility of sample contamination and exposure of hands or personal protective equipment (PPE) to unwanted substances. A flat bottom keeps scoops upright when placed on a flat surface, making them excellent for use as weigh boats. Made from durable, FDA-grade polystyrene plastic, they are safe for food, drug and cosmetic contact. Individually packaged and ready for one time use and disposal. Sterilized by gamma irradiation, lot stamped for accurate lot tracing, and shipped with a Certificate of Sterilization.