Button Cell Battery, Alkaline
Alkaline button cell battery, LR1130, 1.5 V, 50 mAh, 3.05 mm
Catalog # 80086-382
Supplier: Bulbtronics
Fournisseur #: 0033434
Jump to:
Product Details & Documents
For use with certain keyless-entry devices, watches, calculators, cameras, and security electronics.
- Primary cell which is non chargeable
- Alkaline-manganese dioxide(Zn-MnO₂) battery
Spécifications
- Tension:1.5 V
- Capacité:50 mAh
- Désignation:Alkaline button cell battery
- Hauteur:3.05 mm
- Type:LR1130