ColorZip™ Reclosable Storage Bags
: Minigrip
Reclosable polyethylene bags feature easy open/close zipper and external grip strips for simplified handling.
- Single zipper and white write-on block standard for sandwich size
- Double zipper and white write-on block is standard for quart, gallon and jumbo storage and freezer bags
- Double zipper feature provides greater leak-resistance
- Freezer grade is thick and heavy duty
- All bags feature a white write-on block
Large offset lip with external grip ridges for easy handling, even with gloves. Storage bags have improved cold-fracture resistance for short freezing (1 to 2 weeks).
: FDA approved for contact with food.
: Crush-resistant dispenser cartons offer protection in the toughest environments.
Frequently Bought Together