ContactUsLinkComponent
KLEENEX® C-Fold Towels, Kimberly-Clark Professional®
KLEENEX® C-Fold Towels, Kimberly-Clark Professional®
Catalog # CA10199-928
Supplier:  Kimberly-Clark
undefined
KLEENEX® C-Fold Towels, Kimberly-Clark Professional®
Catalog # CA10199-928
Supplier:  Kimberly-Clark
Supplier Number:  01500
Specifications
  • Cat. no.:
    CA10199-928
  • W×L:
    25.4 x 33 cm (10 x 13")
  • Supplier No.:
    01500
Specifications
About this item
Dependable, folded towels provide softness, absorbency, and performance.
Color: white.