Laboratory Bottle, Square, GL Thread, Ace Glass | Avantor
Laboratory Bottle, Square, GL Thread, Ace Glass
Supplier:   Ace Glass
Laboratory Bottle, Square, GL Thread, Ace Glass
Supplier:  Ace Glass
Certificates
Laboratory Bottle, Square, GL Thread, Ace Glass
Laboratory Bottle, Square, GL Thread, Ace Glass
4 Options Available Below
Product Details & Documents
These chemically resistant square bottles are fabricated from borosilicate glass.
They are graduated, and are provided with polypropylene, blue cap, and removable pouring ring that allows drip-free operations. For use up to 140 °C. Not suitable for vacuum or pressure use. Can be hot air sterilized. Height measurement listed is with cap.
Product Family Options
Informations sur le produit
  • Capacité
    Désignation
    Hauteur
    GL Thread Size
    Outer Diameter
    Availability
    Price
  • 100 ml (3.4 oz.)
    Laboratory Square GL Thread Bottle
    109 mm
    32
    50 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $728.11
    $728.11
    Prix promotionnel pour les nouveaux clients
    Case of 10
       $728.11
    Supplier Number: 5560-10
  • 250 ml (8.5 oz.)
    Laboratory Square GL Thread Bottle
    143 mm
    45
    64 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $827.25
    $827.25
    Prix promotionnel pour les nouveaux clients
    Case of 10
       $827.25
    Supplier Number: 5560-15
  • 500 ml (16.9 oz.)
    Laboratory Square GL Thread Bottle
    181 mm
    45
    78 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $1,024.70
    $1,024.70
    Prix promotionnel pour les nouveaux clients
    Case of 10
       $1,024.70
    Supplier Number: 5560-20
  • 1 L (33.8 oz.)
    Laboratory Square GL Thread Bottle
    222 mm
    45
    94 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $1,461.77
    $1,461.77
    Prix promotionnel pour les nouveaux clients
    Case of 10
       $1,461.77
    Supplier Number: 5560-25