Bottle, HDPE, Rectangular, Ace Glass
Bottle, HDPE, Rectangular, Ace Glass
Supplier:   Ace Glass
Bottle, HDPE, Rectangular, Ace Glass
Supplier:  Ace Glass
Certificates
Bottle, HDPE, Rectangular, Ace Glass
Bottle, HDPE, Rectangular, Ace Glass
2 Options Available Below
Product Details & Documents
These rectangular shape wide mouth bottles take less shelf space than standard round bottles.
Sturdy, heavy-wall construction for laboratory use and drug storage. Made of high-density polyethylene (HDPE) with leak-proof screw closures. Well suited for shipping and storing both liquids and dry materials.
Product Family Options
Product Information
  • Capacité
    Désignation
    Taille du bouchon
    Availability
    Price
  • 500 ml (16.9 oz.)
    HDPE Rectangular Bottle
    43
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 12
    Pack of 12 - $258.73
    $258.73
    New Customer Promotional Price
    Log in for availability
    Case of 48
    Case of 48 - $1,034.93
    $1,034.93
    New Customer Promotional Price
    Pack of 12
       $258.73
    Case of 48
       $1,034.93
    Supplier Number: 12431-11
  • 1 L (33.8 oz.)
    HDPE Rectangular Bottle
    53
    Discontinued
    Supplier Number: -
  • 120 ml (4 oz.)
    HDPE Rectangular Bottle
    38
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 12
    Pack of 12 - $114.99
    $114.99
    New Customer Promotional Price
    Log in for availability
    Case of 72
    Case of 72 - $689.93
    $689.93
    New Customer Promotional Price
    Pack of 12
       $114.99
    Case of 72
       $689.93
    Supplier Number: 12431-05