Serum Bottle, Ace Glass
Serum Bottle, Ace Glass
Supplier:   Ace Glass
Serum Bottle, Ace Glass
Supplier:  Ace Glass
Certificates
Serum Bottle, Ace Glass
Serum Bottle, Ace Glass
6 Options Available Below
Product Details & Documents
These borosilicate glass serum bottles meet the requirements of the Pharmacopeia of the United States (USP) for Type I glasses.
  • Repeated sterilization does not affect Type I qualities
  • The mouth Int.Ø and Ext.Ø of bottle is 13×20 mm
Offers maximum protection for delicate injectables and biological materials. They have large mouth openings for ease in filling, emptying, and cleaning.
Product Family Options
Product Information
  • Capacité
    Désignation
    Diameter ×H
    Availability
    Price
  • 5 ml (0.17 oz.)
    Serum Bottle
    23×47 mm
    Log in for availability
    Log in for availability
    Case of 288
    Case of 288 - $766.02
    $766.02
    New Customer Promotional Price
    Case of 288
       $766.02
    Supplier Number: 5530-08
  • 10 ml (0.34 oz.)
    Serum Bottle
    25×54 mm
    Log in for availability
    Log in for availability
    Case of 288
    Case of 288 - $1,065.03
    $1,065.03
    New Customer Promotional Price
    Case of 288
       $1,065.03
    Supplier Number: 5530-10
  • 20 ml (0.6 oz.)
    Serum Bottle
    32×58 mm
    Log in for availability
    Log in for availability
    Case of 288
    Case of 288 - $1,181.68
    $1,181.68
    New Customer Promotional Price
    Case of 288
       $1,181.68
    Supplier Number: 5530-12
  • 30 ml (1 oz.)
    Serum Bottle
    37×63 mm
    Log in for availability
    Log in for availability
    Case of 288
    Case of 288 - $1,601.99
    $1,601.99
    New Customer Promotional Price
    Case of 288
       $1,601.99
    Supplier Number: 5530-14
  • 50 ml (1.7 oz.)
    Serum Bottle
    43×73 mm
    Log in for availability
    Log in for availability
    Case of 288
    Case of 288 - $1,859.97
    $1,859.97
    New Customer Promotional Price
    Case of 288
       $1,859.97
    Supplier Number: 5530-16
  • 60 ml (2 oz.)
    Serum Bottle
    41×91 mm
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $1,256.79
    $1,256.79
    New Customer Promotional Price
    Case of 144
       $1,256.79
    Supplier Number: 5530-18
  • 100 ml (3.4 oz.)
    Serum Bottle
    52×95 mm
    Discontinued
    Supplier Number: 5530-20
  • 125 ml (4.2 oz.)
    Serum Bottle
    54×107 mm
    Discontinued
    Supplier Number: -