Decapper, Ace Glass | Avantor
Decapper, Ace Glass
Supplier:   Ace Glass
Decapper, Ace Glass
Supplier:  Ace Glass
Certificates
Decapper, Ace Glass
Decapper, Ace Glass
2 Options Available Below
Product Details & Documents
This decapper features a new ergonomic design cushioned handle that aids in reducing hand fatigue and provides a higher degree of comfort for the user.
Labeled for quick identification for seal size. The new design removes seals quickly and efficiently. Can be autoclaved.
Product Family Options
Informations sur le produit
  • Désignation
    Seal Size
    Availability
    Price
  • Decapper
    20 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,389.10
    $1,389.10
    Prix promotionnel pour les nouveaux clients
    Each
       $1,389.10
    Supplier Number: 5535-07
  • Decapper
    11 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,490.00
    $2,490.00
    Prix promotionnel pour les nouveaux clients
    Each
       $2,490.00
    Supplier Number: 5535-03