NMC Webbed Barrier Tape
Magenta/yellow, 5.08 cm x 45.72 m (2" x 50 yds)
Catalog # 94004-420
Supplier: ACCUFORM MANUFACTURING, INC
Supplier #: BT5MY
Jump to:
Product Details & Documents
Ideal for permanent outdoor marking applications
Made from woven polypropylene. Chemical resistant, weather proof, faced resistant. Tensile strength is in excess of 220lbs.
Spécifications
- Color:Magenta/yellow
- Taille:5.08 cm x 45.72 m (2" x 50 yds)