Bottle, Threaded Neck, Ace Glass
 :   Ace Glass
Bottle, Threaded Neck, Ace Glass
 :  Ace Glass
Bottle, Threaded Neck, Ace Glass
Bottle, Threaded Neck, Ace Glass
This is a borosilicate glass bottle designed for the storage of solutions.
Bottles have a GL120 external thread finish neck with large 106 mm (4.2") Int.Ø opening. The 9.5 and 13.25 liter sizes have a conventional bottle shape. The 19 and 45.5 liter sizes are similar in design to a carboy.
 : A glass-filled PTFE cap with CAPFE (PTFE encapsulated silicone) O-ring may be purchased separately. Ask for Order Code 89059-794.
Product Family Options
  • Capacité
    Désignation
    Hauteur
    Ø
  • 13.2 L (3.5 gal.)
    Threaded Neck, Solution Bottle
    380 mm
    238 mm
    Each
    Each - $1,477.76
    $1,477.76
    Each
       $1,477.76
     : 4048-13
  • 19 L (5 gal.)
    Threaded Neck, Solution Bottle
    480 mm
    292 mm
    Each
    Each - $1,721.09
    $1,721.09
    Each
       $1,721.09
     : 4048-19
  • 45.5 L (12 gal.)
    Threaded Neck, Solution Bottle
    520 mm
    406 mm
    Each
    Each - $3,194.30
    $3,194.30
    Each
       $3,194.30
     : 4048-45
  • 9.5 L (2.5 gal.)
    Threaded Neck, Solution Bottle
    460 mm
    187 mm
    Each
    Each - $1,268.30
    $1,268.30
    Each
       $1,268.30
     : 4048-09