ContactUsLinkComponent
Beakers, Jacketed, Ace Glass Incorporated
Catalogue # 89051-596
Fournisseur:  Ace Glass
Fournisseur #: 5340-15
Product available on GSA Advantage®
Small Business Enterprise
undefined
Beakers, Jacketed, Ace Glass Incorporated
Catalogue # 89051-596
Fournisseur:  Ace Glass
Supplier Number:  5340-03
Fournisseur #: 5340-15
Product available on GSA Advantage®
Small Business Enterprise
S'identifier pour la disponibilité
Select an Option
81 mm (I.D.)
Select an Option
600 ml (20 oz.)
Select an Option
Jacketed Beaker
Select an Option
152 mm
Select an Option
89051-596
Select an Option
3/8 (D) Hose Connection
Select an Option
5340-15
À propos de cet article
This jacketed beaker has one upper and one lower hose connection on opposite sides
Does not have a pouring spout. Fabricated from borosilicate glass.
Frequently Bought Together