VWR® Hazmat Spill Response Kit
Catalog # 89168-998
Supplier: VWR International
Specifications
- Cat. no.:89168-998
- Absorbency:34.1 L (9 gal.) of acids, caustics, bases
Specifications
About this item
Hazmat spill response kit includes 20 pads, three socks, one pair nitrile gloves, one pair safety goggles, and one water-resistant yellow bag with tie.
- Includes 20 pads, three socks, one pair nitrile gloves, one pair safety goggles, and one water-resistant yellow bag with tie
- Supports compliance with 40 CFR 263.30(a) safety standard