NMC Valve Wheel Lockouts
NMC Valve Wheel Lockouts
Certificates
NMC Valve Wheel Lockouts
NMC Valve Wheel Lockouts
5 Options Available Below
Product Details & Documents
Lockouts consist of two flattened half moons that completely cover the valve wheels from 1" to 14" in diameter
Made from strong Polypro plastic, fully dielectric. Highly resistant to cracking and abrasion. Resistant to extreme temperature changes 50⁰F
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Fits 2.54 cm (1") To 6.35 cm (2.5") Dia.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $98.92
    $98.92
    Each
       $98.92
    Supplier Number: VL02R
  • Fits 6.35 cm (2.5") To 12.7 cm (5") Dia.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $163.85
    $163.85
    Each
       $163.85
    Supplier Number: VL04R
  • Fits 12.7 cm (5") To 16.51 cm (6.5") Dia.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $199.34
    $199.34
    Each
       $199.34
    Supplier Number: VL06R
  • Fits 16.51 cm (6.5") To 25.4 cm (10") Dia.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $248.90
    $248.90
    Each
       $248.90
    Supplier Number: VL09R
  • Fits 25.4 cm (10") To 35.56 cm (14") Dia.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $481.30
    $481.30
    Each
       $481.30
    Supplier Number: VL13R