ContactUsLinkComponent
Beakers, Griffin Low Form, Ace Glass Incorporated
  89085-386
 :  Ace Glass
 : 12420-22
Product available on GSA Advantage®
Small Business Enterprise
undefined
Beakers, Griffin Low Form, Ace Glass Incorporated
  89085-386
 :  Ace Glass
 : 12420-22
Product available on GSA Advantage®
Small Business Enterprise
Select an Option
4 L (135 oz.)
Select an Option
Griffin Low Form Beaker
Select an Option
89085-386
Select an Option
12420-22
These Griffin-style polypropylene beakers are for general laboratory use
  • Autoclavable
  • Approximate raised volume scales
  • Tapered walls for safe handling and convenient stacking
Combines a “no-drip” pouring spout with unbreakability and maximum translucency.
 : Lids are not included.
Frequently Bought Together