PC Link Labels, Brady
PC Link Labels, Brady
Supplier:   Avantor
PC Link Labels, Brady
Supplier:  Avantor
Certificates
PC Link Labels, Brady
PC Link Labels, Brady
3 Options Available Below
Product Details & Documents
Labels for the TLS 2200® and TLS PC Link™ Thermal transfer portable printers.
  • Self-laminating
  • Self-extinguishing
  • Excellent water and oil resistance
  • Excellent abrasion and smudge resistance
B-427 is an excellent material for wire and cable identification and is not recommended for use in harsh organic solvents such as methyl ethyl ketone, acetone, or 1,1,1-trichloroethane.
This material has good clarity and conformability.
Certifications: RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC.
Product Family Options
Product Information
  • Désignation
    Matière
    Color
    Pour l'utilisation avec
    Dimensions
    Availability
    Price
  • Tls printer labels B-427 MAT´L
    Self-laminating Vinyl
    White/translucent
    TLS 2200, TLS-PC Link, BMP71 printers
    6.4×27.9 mm (0.25×1.1")
    Log in for availability
    Log in for availability
    Each
    Each - $97.84
    $97.84
    Each
       $97.84
  • Tls printer labels B-427 MAT´L
    Self-laminating Vinyl
    White
    TLS 2200, TLS-PC Link, BMP71 printers
    19.1×38.1 mm (0.75×1.5")
    Log in for availability
    Log in for availability
    Each
    Each - $72.26
    $72.26
    Each
       $72.26
  • Tls printer labels B-427 MAT´L
    Self-laminating Vinyl
    White
    TLS 2200, TLS-PC Link, BMP71 printers
    25.4×48.3 mm (1×1.9")
    Log in for availability
    Log in for availability
    Each
    Each - $97.84
    $97.84
    Each
       $97.84