PC Link Labels, Brady | Avantor
PC Link Labels, Brady
Supplier:   Avantor
PC Link Labels, Brady
Supplier:  Avantor
Certificates
PC Link Labels, Brady
PC Link Labels, Brady
2 Options Available Below
Product Details & Documents
B-439 is a colored vinyl film with an acrylic pressure sensitive adhesive and a topcoat specifically formulated for thermal transfer printing.
Vinyl colors include the following: Silver, Gold, Red, Purple, Yellow, Orange, Green, Light Blue, Black, and White. B-439 is designed for use in ambient conditions with limited solvent exposure. Recommended ribbons are Brady Series R4900 and R6000 black ribbons and R4400W white ribbon.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Color
    Pour l'utilisation avec
    Width
    Roll Length
    Availability
    Price
  • Tls printer labels B-439 MAT´L
    Permanent Vinyl
    Orange
    TLS 2200, TLS-PC Link, BMP71 printers
    1"
    15.24 m (50 ft.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $128.96
    $128.96
    Each
       $128.96
  • Tls printer labels B-439 MAT´L
    Permanent Vinyl
    Yellow
    TLS 2200, TLS-PC Link, BMP71 printers
    1"
    15.24 m (50 ft.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $128.96
    $128.96
    Each
       $128.96