Scribes, Double Ended, Excelta Corp®
Supplier:   Excelta
Scribes, Double Ended, Excelta Corp®
Supplier:  Excelta
Certificates
Scribes, Double Ended, Excelta Corp®
Scribes, Double Ended, Excelta Corp®
2 Options Available Below
Product Details & Documents
These double ended scribers are made of stainless steel, with hardened points.
  • Double ended for expanded utility
  • Cleanroom safe, stainless steel
They feature a knurled, finger-grip handle.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Point Size
    Availability
    Price
  • Stainless steel AISI 440 scribe, double ended, 3 star quality, 90° point angle
    212.5 mm (8.5")
    0.38×0.25 mm (0.015×0.010")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $20.99
    $20.99
    Each
       $20.99
    Supplier Number: 477
  • Stainless steel AISI 440 scribe, double ended, 3 star quality
    187.5 mm (7.5")
    0.38×0.25 mm (0.015×0.010")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $29.06
    $29.06
    Each
       $29.06
    Supplier Number: 478