ContactUsLinkComponent
Plier, Roll Pin Insertion-Extraction, Excelta
  63043-008
 :  Excelta
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Plier, Roll Pin Insertion-Extraction, Excelta
  63043-008
 :  Excelta
 :  900-100X
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
  • Description:
    Five Star™ Plier
  • Material:
    Carbon steel
  • Cat. no.:
    63043-008
  • Tip Style:
    Tool without jaws
  • Overall Length:
    16.5 cm (6½")
  • Supplier No.:
    900-100X
These tools are designed to insert or extract the card key roll pin, thus reducing production time.