Probes, Fine Tip, Stainless Steel, Excelta Corp®
Supplier: Excelta
Certificates
About this item
These 0.01" probes are ideal for fine work.
- Cleanroom safe, stainless steel
- 3" overall length for work under a scope
This series three star fine straight tip stainless steel probe has 10 mil diameter tip for precise work. They are offered with a standard 6" overall length and also in a 3" version.
Frequently Bought Together