Nalgene® LDPE Narrow Mouth Packaging Bottles, with PP Closures, Bulk Pack, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
À propos de cet article
Nalgene low density polyethylene narrow mouth packaging bottles have a robust construction for packaging use and are featured with polypropylene closures.
These packaging bottles have excellent impact chemical resistance and clarity. They are constructed of the ideal material for trace metal analysis
Spécifications
- Catalog No:
- 16059-540
- 16059-544
- 16059-546
- Environmentally Preferable:
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)