About this item
Lessen environmental impact and increase contributions to sustainability goals with these PLA (polylactic acid) resin sampling spoons.
- PLA resin construction
- Earth-friendly with no petroleum-based materials
- Long handle helps prevent contact with sample material
- Nonsterile
The production of 100% plant-based PLA contributes less greenhouse gasses and uses less nonrenewable energy from source to resin than its petroleum-based counterparts, and the plants used are an annually renewable resource.
Disposal options include industrial composting for biodegradability, incineration, recycling, and landfill. Safe for food, drug, and cosmetic contact, these FDA-grade spoons are comparable in strength and durability to polystyrene, polypropylene, and other plastic resin sampling spoons.
A long handle helps prevent personal contact with sampled material when reaching into bottles, jars, and other confined areas. The rim of each bowl has a flat edge to assist in collecting and measuring small amounts with accuracy. Use a separate spoon for each material sampled to avoid cross contamination. Nonsterile.
Specifications
- Catalog No:
- 89218-788
- 89218-790
- 89218-786
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing