ContactUsLinkComponent
Nalgene® Autoclavable Septum Closure, PP closure, Thermoplastic Elastomer Septum, Thermo Scientific
Catalog # 16210-081
undefined
Nalgene® Autoclavable Septum Closure, PP closure, Thermoplastic Elastomer Septum, Thermo Scientific
Catalog # 16210-081
Supplier Number:  DS2168-0384
About this item
Thermo scientific nalgene autoclavable septum closures allow aseptic injection of reagent or sample withdrawal without compromising sterility or integrity of contents.
Suitable for use with any bottle or container with 38-430 neck, including Nalgene culture vessels.