SANI-STICK Sampling Kits, Labplas
SANI-STICK Sampling Kits, Labplas
Supplier:   Labplas
SANI-STICK Sampling Kits, Labplas
Supplier:  Labplas
Certificates
SANI-STICK Sampling Kits, Labplas
SANI-STICK Sampling Kits, Labplas
4 Options Available Below
Product Details & Documents
The SANI-STICK is designed to collect samples to detect the presence of microbiological contaminations on any surface.
  • Allows swabbing of hard-to-reach areas
  • Convenient and east to use
  • Helps eliminate several time -consuming steps
These sponges on sticks are widely used in the food industry applications such as quality assurance procedures (QA/QC).
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    W×L
    Availability
    Price
  • SANI-STICK kit with wet sponge on handle with letheen broth
    600 ml
    140×279 mm (5.5×11")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $1,396.25
    $1,396.25
    Case of 100
       $1,396.25
    Supplier Number: KSS-67310-LT
  • SANI-STICK kit with wet sponge on handle with buffered peptone water
    600 ml
    140×279 mm (5.5×11")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $1,772.21
    $1,772.21
    Case of 100
       $1,772.21
    Supplier Number: KSS-67310-BPW
  • SANI-STICK kit with wet sponge on handle with neutralizing buffer
    600 ml
    140×279 mm (5.5×11")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $1,505.85
    $1,505.85
    Case of 100
       $1,505.85
    Supplier Number: KSS-67310-NE-VW
  • SANI-STICK kit with wet sponge on handle with DE-neutralizing buffer
    600 ml
    140×279 mm (5.5×11")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $1,644.72
    $1,644.72
    Case of 100
       $1,644.72
    Supplier Number: KSS-67310-DE-VW