Dispenser with 8 Compartments, S-Curve Technologies
Eight compartments to organize and store small items.
- 1/4″ clear acrylic or 1/4″ clear PETG material
- Open top for easy access to product
- Flame polished edges for a smooth finish
- Keyhole standoffs on back for wall mounting
Wall mountable: no.
- Catalog No:
- 89208-626
- 89208-624
- Material:
- PETG
- Acrylic
- No. of compartments:
- 8
- 8
- Dispenser type:
- Bench top
- Bench top
Frequently Bought Together