ART® Solvent Safe Micropoint Pipette Tips, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
À propos de cet article
ART specialty pipette tips are available in extended length to prevent contamination by keeping the pipette shaft from entering the sample vessel, gel loading for loading acrylamide or agarose gels, wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials, and Solvent Safe for acids, bases and aggressive organic solvents.
Spécifications
- Catalog No:
- 72830-008
- Environmentally Preferable:
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)