Molecular BioProducts pipette tips are designed to fit AP96, FX/NX, 1000, 2000, and 3000 liquid handling workstations.
- Designed for compatibility and performance
- Variety of sizes and styles available
- Lot-tested using 15-point program
BioRobotix® robotic pipette tips are available in low retention for critical samples and in extended length for increased volume and the ability to reach into deep well plates.
Genomic tips are excellent for pipetting of more viscous samples. The tips have an orifice that is up to 260% larger than the standard workstation tip orifice. This provides more precision when pipetting viscous tissue samples, such as blood, and prevents shearing of genomic DNA during sample preparation and processing.
Automation pipette tips for the 1000 and 2000 automated liquid handling systems feature precise design and optimal fit for accuracy. Wide selection of tips in both standard and liquid-level sensing carbon black-conductive polypropylene are available. Automation pipette tips for the 1000 and 2000 are certified RNase-, DNase-, and pyrogen-free.
Automation pipette tips for the FX and NX automated liquid handling systems have been designed, validated, and tested on the FX workstation to ensure equal performance without the need to modify your existing protocols and programs. The low percentage Coefficient of Variation (%CV) ensures product consistency and performance from every lot. Automation pipette tips for the FX and NX are certified RNase, DNase, DNA, endotoxin, bioburden, and pyrogen free.
The pipette tips, racks and lids reduce static charges so tips pick up straight and eject perfectly each time. These tips are designed with SoftFit™ feature that enhances workstation performance and longevity by giving a more secure fit with reliable, consistent loading and ejecting. Filter tips utilize the ART barrier for guranteed protection against cross-contamination.
- Catalog No:
- 72830-412
- 89025-880
- 72830-408
- 89025-876
- 89025-878
- 72830-290
- 72830-528
- 72830-353
- 72830-210
- 76457-542
- 72830-518
- 46300-372
- 46300-370
- 72830-218
- 46300-362
- 72830-216
- 72830-212
- 53226-018
- 53226-016
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)