DrumQuik PRO Series Drum and Tote, Dip Tube and Connection Technology, Colder Products Company
Fournisseur: Colder Products
Certificats
À propos de cet article
Engineered to provide safe, secure container change outs with a quick connect and disconnect technology and integrated dip tube
Materials of construction include polypropylene or PVDF, HDPE, 316 stainless steel or an all plastic spring, EPDM, FKM or FFKM o-ring seals.
Convert and open dispensing system into a closed dispensing system with the use of any common pump.
Ordering information: Accessories not included
Frequently Bought Together