The Hamilton line of pipettes puts a priority on addressing the issues of hand strain, fatigue and other injuries.
- Slim ejection sleeve is made from a high strength, chemically resistant polymer
- Slim design allows the pipette to reach to the bottom of narrow tubes without the need for extended length tips
SoftGrip™ adjustable volume pipettes are available in 0.2 µl to 1 ml. The isolated volume adjustment makes it so there's no need for a locking mechanism to prevent accidental volume adjustment.
The new generation of Hamilton pipettes has a dual indicator that improves the volume adjustment accuracy by removing the parallax errors.
SoftGrip™ pipettes have a universal calibration key that reduces the need for costly calibration services. Calibration requires only minutes to perform and eliminates the downtime and cost of alternative calibration methods.
The SoftGrip™ tip nozzle ensures a proper fit with universal pipette tips. Unique color coding and labeling clearly indicates the working ranges of the adjustable volume pipettes.
Our products set new standards for quality, comfort and precision.