SoftGrip™ Single-Channel Fixed Volume Manual Pipettors, Hamilton Company
Supplier: Hamilton
Certificates
About this item
The Hamilton line of pipettors puts a priority on addressing the issues of hand strain, fatigue, and other injuries.
- Slim ejection sleeve is made from a high strength, chemically resistant polymer
- Slim design allows the pipette to reach to the bottom of narrow tubes without the need for extended length tips
The SoftGrip™ fixed volume pipettors are available in volume ranges from 5 µl to 1ml.
SoftGrip™ pipettors have a universal calibration key that reduces the need for costly calibration services. Calibration requires only minutes to perform and eliminates the downtime and cost of alternative calibration methods.
The SoftGrip™ tip nozzle ensures a proper fit with universal pipettor tips. Unique color coding and labeling clearly indicates the volume of the pipettor.
Hamilton products set new standards for quality, comfort and precision.
Frequently Bought Together