FlexStation® 3 Multimode Plate Reader, Molecular Devices
FlexStation® 3 Multimode Plate Reader, Molecular Devices
Supplier:   Molecular Devices
FlexStation® 3 Multimode Plate Reader, Molecular Devices
Supplier:  Molecular Devices
Certificates
FlexStation® 3 Multimode Plate Reader, Molecular Devices
FlexStation® 3 Multimode Plate Reader, Molecular Devices
3 Options Available Below
Product Details & Documents
FlexStation® 3 Benchtop Multi-Mode Microplate Reader combines Molecular Devices’ SpectraMax® M5e Multi-Mode Microplate Reader performance with an integrated 8- or 16-channel pipettor into one compact benchtop reader.
  • Integrated pipettor
  • High-efficiency optics
  • Several configurations
The FlexStation 3 Microplate Reader high-efficiency monochromator optics are tunable for absorbance, fluorescence intensity, fluorescence polarization, and time-resolved fluorescence assays
Users have several choices when configuring the FlexStation 3 Microplate Reader. Users can choose from 96-well pipetting, 384-well pipetting, or both. Once installed, the pipettors can be changed by the user with no tools in under 5 minutes.
Using an 8- or 16-channel pipettor, the FlexStation 3 Microplate Reader offers added assay flexibility over dispenser-based systems by directly transferring reagents from 96- or 384- well source plates to a read plate, one column at a time. In addition, users can define individual reagents and concentrations to be delivered to each column. The direct transfer capability reduces reagent consumption and allows more assay conditions to be explored in a single microplate, making the system more amenable to agonist and antagonist assay formats.
It also has a dedicated photomultiplier tube for luminescence assays.
Ordering information: Includes current version of SoftMax Pro Software. Additional models are available. Additional accessories are available including the µMax Low Volume Plate, standard validation tools, and 21 CFR Part 11 Compliance tools. An 8-Channel and/or 16-Channel head must be selected at the time of purchase. Standard 12-month warranty included. Purchase of Additional 1 Year Service Plan includes: MRC Essential Service Plan, including parts, labor, and shipping/handling costs for emergency repairs for one year at the Molecular Devices Repair Center. Priority repair scheduling, along with unlimited technical support and loaner instrument (if available), are provided with this service option.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • FlexStation® 3 Microplate Reader
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each
    Supplier Number: FLEX3
  • FlexStation® 3 Microplate Reader with 1-Year Service Contract
    Discontinued
    Supplier Number: -
  • FlexStation® 3 8-channel head
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $18,117.43
    $18,117.43
    Each
       $18,117.43
    Supplier Number: 0200-6182
  • FlexStation® 3 8-channel head
    Discontinued
    Supplier Number: 0200-6182
  • FlexStation® 3 16-channel head
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $36,235.43
    $36,235.43
    Each
       $36,235.43
    Supplier Number: 0200-6183
  • FlexStation® 3 16-channel head
    Discontinued
    Supplier Number: 0200-6183
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Flexstation Pipet Tips (Black), 96-Well, 200 µL
    Flexstation Pipet Tips (Black), 96-Well, 200 µL
    CA89212-408
    Log in pour la disponibilité
    Case of 10 - $249.21