Condenser, Allihn
Supplier: Ace Glass
Jump to:
À propos de cet article
Bulb type Allihn condenser, approximately one bulb per every 50 mm, with standard taper inner joint at bottom and reinforced standard taper outer joint at top
Fabricated from borosilicate glass. Size D glass hose connections, for use with 3/8-inch I.D. tubing.
12 Options Available Below
Product Details & Documents
Bulb type Allihn condenser, approximately one bulb per every 50 mm, with standard taper inner joint at bottom and reinforced standard taper outer joint at top
Fabricated from borosilicate glass. Size D glass hose connections, for use with 3/8-inch I.D. tubing.
Product Family Options
Informations sur le produit
- DésignationST JointJacket LengthAvailabilityPrice
- Supplier Number: 5945-12
- Supplier Number: 5945-05
- Supplier Number: 5945-15
- Supplier Number: 5945-16
- Supplier Number: 5945-17
- Supplier Number: 5945-24
- Supplier Number: 5945-25
- Supplier Number: 5945-26
- Supplier Number: 5945-27
- Supplier Number: 5945-34
- Supplier Number: 5945-14
- Supplier Number: 5945-13