Condenser, Allihn, Jacketed
 :   Ace Glass
Condenser, Allihn, Jacketed
 :  Ace Glass
Condenser, Allihn, Jacketed
Condenser, Allihn, Jacketed
Bulb type Allihn condenser, approximately one bulb per every 50 mm, with 24/40 standard taper inner joint at bottom, open tube at the top
Fabricated from borosilicate glass. Use with 3/8-inch I.D. tubing, size D hose connections.
Product Family Options
  • Désignation
    Jacket Length
  • Condenser, Alihn, Jacketed
    200 mm
    Each
    Each - $381.99
    $381.99
    Each
       $381.99
     : 5941-12
  • Condenser, Alihn, Jacketed
    300 mm
    Each
    Each - $414.78
    $414.78
    Each
       $414.78
     : 5941-14