Split Stoppers, Labconco®
Split Stoppers, Labconco®
Supplier:   Labconco
Split Stoppers, Labconco®
Supplier:  Labconco
Certificates
Split Stoppers, Labconco®
2 Options Available Below
Product Details & Documents
Split Stoppers
Package of 100. Rubber stopper, when crimped with an aluminum seal, tightly seals Serum Bottle for lyophilization applications. For use with FreeZone Freeze Dry Systems. Stoppers are for serum bottles with 20 mm corkage or 13 mm corkage, as indicated.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Split Stopper, 13 mm Corkage
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $71.80
    $71.80
    Pack of 100
       $71.80
    Supplier Number: 7576010
  • Split Stopper, 20 mm Corkage
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $77.50
    $77.50
    Pack of 100
       $77.50
    Minimum order quantity is 1
    Supplier Number: 7576210