Replacement HEPA Filter for 3' Certain 3' Enclosures, Labconco®
Catalog # 26673-442
Supplier: Labconco
Product available on GSA Advantage®
Specifications
- Description:Replacement HEPA Filter
- Cat. no.:26673-442
- Supplier No.:3707901
Specifications
About this item
Replacement HEPA Filter, 99.99% efficient HEPA Filter
For use with 3' Purifier Class I and HEPA Filtered Enclosures, 3' Vertical Clean Benches; 3' and 6' XPert Filtered Systems and Stations; PuriCare Vertical Flow Stations and Bedding Disposal Stations.