XPert® TXE™ Equipment Enclosures, Labconco
Fournisseur: Labconco
Certificats
À propos de cet article
XPert TXE Equipment Enclosures are designed to address the needs of pharmaceutical research and quality control laboratories
These vented equipment enclosures may be ducted to the outside or connected to the FilterMate Portable Exhauster for HEPA and/or carbon filtered exhaust back into the laboratory. Specialized base stands are also available (sold separately).
It provides a safe, ventilated space for keeping powder-generating apparatus contained. Its tall 42" interior height accommodates equipment such as pill presses, grinders, crushers and sieves. HEPA-filtered air is returned to the laboratory.
Frequently Bought Together