About this item
G-Biosciences' avidin is a glycoprotein with approximately 10% of its total mass comes from carbohydrates
- Specific activity 15.3 U/mg protein
Avidin is a basic protein with an isoelectric pH of 10 to 10.5 and is readily soluble in aqueous buffers containing a wide range of salt, pH, temperature and other laboratory agents. This wide range of tolerance makes avidin suitable for a wide variety of analytical applications.
G-Biosciences' affinity purified avidin is ideal for estimation of biotin incorporation and other applications.
Avidin has a molecular weight of 67 kDa and contains four identical 128 amino acid subunits that each have a single biotin binding domain.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.