Precision Screwdriver, Aven
Precision Screwdriver, Aven
Supplier:   Aven Tools
Precision Screwdriver, Aven
Supplier:  Aven Tools
Certificates
Precision Screwdriver, Aven
Precision Screwdriver, Aven
2 Options Available Below
Product Details & Documents
Specifically designed for electronics
Product Family Options
Informations sur le produit
  • Désignation
    Blade Length
    Tip Size
    Availability
    Price
  • Slotted Driver
    50 mm (2")
    1.6 mm
    Discontinued
    Supplier Number: 13901
  • Slotted Driver
    50 mm (2")
    2.0 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $21.21
    $21.21
    Each
       $21.21
    Supplier Number: 13902
  • Slotted Driver
    50 mm (2")
    2.4 mm
    Discontinued
    Supplier Number: 13903
  • Slotted Driver
    50 mm (2")
    3.0 mm
    Discontinued
    Supplier Number: 13904
  • Phillips Driver
    50 mm (2")
    No. 000
    Discontinued
    Supplier Number: 13910
  • Phillips Driver
    50 mm (2")
    No. 00
    Discontinued
    Supplier Number: 13911
  • Phillips Driver
    50 mm (2")
    No. 1
    Discontinued
    Supplier Number: 13913
  • Phillips Driver
    102 mm (4")
    No. 1
    Discontinued
    Supplier Number: 13915
  • Torx Driver
    50 mm (2")
    T6
    Discontinued
    Supplier Number: -
  • Torx & Tamper-Proof Driver
    50 mm (2")
    T8
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $21.94
    $21.94
    Each
       $21.94
    Supplier Number: 13923
  • Nut Driver
    50 mm (2")
    M3.0
    Discontinued
    Supplier Number: -
  • Nut Driver
    50 mm (2")
    M4.0
    Discontinued
    Supplier Number: 13932
  • Nut Driver
    50 mm (2")
    M4.5
    Discontinued
    Supplier Number: 13933