ART® Barrier Ultra Micro Tips with MicroPoint Design, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
À propos de cet article
Molecular Bioproducts ART® pipette tips are pre-sterilized and precision molded with the ART® self-sealing barrier.
They are guaranteed to prevent carryover contamination during aspiration and delivery of samples. Ideally suited for low volume applications in genetic studies, forensics, PCR, and radioisotope sampling.
Spécifications
- Catalog No:
- 89082-106
- 89082-080
- 89082-078
- 89082-092
- 89082-070
- 53509-500
- 89082-090
- 89082-068
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)