ContactUsLinkComponent
GELPLATE-clean™ for Cleaning Electrophoresis Gel Plates, G-Biosciences
GELPLATE-clean™ for Cleaning Electrophoresis Gel Plates, G-Biosciences
Catalog # CA82021-306
Supplier:  G-Biosciences
undefined
GELPLATE-clean™ for Cleaning Electrophoresis Gel Plates, G-Biosciences
Catalog # CA82021-306
Supplier:  G-Biosciences
Supplier Number:  786-140
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    GELPLATE-clean™ Spray
  • Size:
    2 x 250 mL
  • Cat. no.:
    CA82021-306
  • Supplier No.:
    786-140
Specifications
About this item
G-Biosciences' GELPLATE-clean™ has been specifically formulated for cleaning electrophoresis gel plates. Clean your electrophoresis plates with GELPLATE-clean™ to prevent poor electrophoretic band migration, band distortions, and poor image development.
G-Biosciences' GELPLATE-clean™ removes proteins, nucleic acids, fats, lipids, radioisotopes and other contaminants from electrophoresis gel plates. The solution is suitable for both sequencing and protein electrophoresis gel plates. With GELPLATE-clean™, simply spray your electrophoresis plates and then wipe clean.
Ordering information: Refill solutions are offered separately. Accessories available.
Packaging: Supplied in two easy-to-use 250 mL spray bottles.