To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:500 bp DNA Ladder
- Size:100 loads
- Cat. no.:CA95057-692
- Supplier No.:786-462
Specifications
About this item
G-Biosciences' 1.0 kb DNA ladder is a ready-to-use preparation that consists of 15 DNA fragments ranging from 100 bp to 10 kb
G-Biosciences' 500 bp DNA ladder is a ready-to-use preparation that consists of 10 DNA fragments ranging from 500 bp to 5.0 kb. The recommended loading is 5 μL DNA marker per lane. The 500 bp ladder can be used for approximating the mass of DNA in comparably intense samples of similar size.
G-Biosciences' 100 bp DNA marker is a ready-to-use preparation that consists of 14 fragments, ranging form 100bp to 3,000 bp. The recommended loading is 5 μL DNA marker per lane. The 100 bp ladder cannot be used for precise quantification of DNA mass, but can be used for approximating the mass of DNA in comparably intense samples of similar size.
G-Biosciences' 50 bp DNA marker is a ready-to-use preparation that consists of 13 fragments, ranging form 50 bp to 1,000 bp. The recommended loading is 5 μL DNA marker per lane. The 50 bp ladder cannot be used for precise quantification of DNA mass, but can be used for approximating the mass of DNA in comparably intense samples of similar size.
The recommended loading is 5 μL DNA marker per lane. The 1.0 kb ladder can be used for approximating the mass of DNA in comparably intense samples of similar size.